Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005113.3 | NP_005104.3 | 2 | 463 | ||
XM_011537420.2 | XP_011535722.1 | 2 | 463 | |||
GenBank mRNA | AF085199.1 | - | 2 | 338 | ||
AJ132949.1 | - | 1 | 207 | |||
AK293077.1 | - | 2 | 366 | |||
AK303789.1 | - | 2 | 369 | |||
AL702150.1 | - | 2 | 391 | |||
AY644768.1 | - | 1 | 243 |