Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOPEY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320714.1 | NP_001307643.1 | 20 | 4956 | ||
NM_005128.3 | NP_005119.2 | 20 | 4847 | |||
XM_017028509.1 | XP_016883998.1 | 17 | 4350 | |||
GenBank mRNA | AB023150.2 | - | 17 | 4316 | ||
AJ237839.2 | - | 20 | 4853 | |||
AK025095.1 | - | 2 | 448 | |||
BC012791.2 | - | 2 | 489 |