Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173533.1 | NP_001167004.1 | 7, 8 | 1286 | ||
NM_001173534.1 | NP_001167005.1 | 7, 8 | 1277 | |||
NM_001184781.1 | NP_001171710.1 | 8, 9 | 1400 | |||
NM_005137.2 | NP_005128.1 | 8, 9 | 1409 | |||
NR_033674.1 | 7, 8 | 1294 | ||||
GenBank mRNA | AK291670.1 | - | 8, 9 | 1367 | ||
AK295697.1 | - | 7, 8 | 1252 | |||
AK296337.1 | - | 7, 8 | 1235 | |||
AK303130.1 | - | 8, 9 | 1319 | |||
AK304382.1 | - | 7, 8 | 1244 | |||
BC020339.1 | - | 8, 9 | 1356 | |||
BC032430.1 | - | 8, 9 | 1345 | 5243412 | ||
BC035425.1 | - | 8, 9 | 1356 | |||
BC040500.1 | - | 8, 9 | 1404 | 5272271 | ||
CR456433.1 | - | 8, 9 | 1181 | |||
CR936871.1 | - | 9, 10 | 1744 | |||
D79985.1 | - | 8, 9 | 1346 | |||
L77560.1 | - | 8, 9 | 1310 | |||
X83545.1 | - | 8, 9 | 1310 | |||
X84076.1 | - | 8, 9 | 1307 |