Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135729.1 | NP_001129201.1 | 8, 9 | 958 | ||
NM_001135730.1 | NP_001129202.1 | 7, 8 | 896 | |||
NM_001135732.1 | NP_001129204.1 | 8, 9 | 1031 | |||
NM_005488.2 | NP_005479.1 | 8, 9 | 1031 | |||
NR_024194.1 | 8, 9 | 1060 | ||||
NR_024195.1 | 7, 8 | 981 | ||||
XM_011529818.2 | XP_011528120.1 | 9, 10 | 1045 | |||
XM_011529820.2 | XP_011528122.1 | 7, 8 | 896 | |||
XM_017028529.1 | XP_016884018.1 | 9, 10 | 1045 | |||
XM_017028530.1 | XP_016884019.1 | 7, 8 | 896 | |||
GenBank mRNA | AJ006973.1 | - | 8, 9 | 967 | ||
AK026576.1 | - | 8, 9 | 984 | |||
AK097066.1 | - | 6, 7 | 542 | |||
AK293694.1 | - | 8, 9 | 958 | |||
AK295918.1 | - | 7, 8 | 891 | |||
AK296670.1 | - | 8, 9 | 968 | |||
AK297827.1 | - | 7, 8 | 810 | |||
AK314522.1 | - | 8, 9 | 940 | |||
BC040006.1 | - | 8, 9 | 924 | |||
BC046151.1 | - | 8, 9 | 909 | 6470390 | ||
CR456595.1 | - | 8, 9 | 927 |