Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177465.2 | NP_001170936.1 | 3 | 891 | ||
NM_001177466.2 | NP_001170937.1 | 2 | 638 | |||
NM_005491.4 | NP_005482.2 | 3 | 713 | |||
XM_006724802.3 | XP_006724865.1 | 5 | 1061 | |||
XM_006724803.3 | XP_006724866.1 | 4 | 986 | |||
XM_011531092.2 | XP_011529394.1 | 5 | 1061 | |||
XM_011531093.2 | XP_011529395.1 | 6 | 1191 | |||
XM_011531095.2 | XP_011529397.1 | 5 | 1061 | |||
XM_017029188.1 | XP_016884677.1 | 5 | 933 | |||
XM_017029189.1 | XP_016884678.1 | 5 | 934 | |||
GenBank mRNA | AK294478.1 | - | 3 | 750 | ||
AK302612.1 | - | 4 | 755 | |||
AK315217.1 | - | 3 | 586 | |||
BC136324.1 | - | 2 | 623 | 9020311 | ||
U46023.1 | - | 3 | 714 |