Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278556.1 | NP_001265485.1 | 4 | 349 | ||
NM_001287222.1 | NP_001274151.1 | 4 | 349 | |||
GenBank mRNA | AF004561.1 | - | 3 | 198 | ||
AF006086.1 | - | 4 | 223 | |||
AK297309.1 | - | 4 | 280 | |||
AL357196.1 | - | 4 | 259 | |||
BC067747.1 | - | 4 | 255 | 5302338 | ||
BC078162.1 | - | 4 | 232 | 6148231 | ||
BG620669.1 | - | 4 | 267 | |||
BG723870.1 | - | 4 | 313 | |||
CR407667.1 | - | 3 | 198 | |||
DQ328220.1 | - | 4 | 279 |