Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168412.1 | NP_001161884.1 | 3 | 464 | ||
NM_005724.5 | NP_005715.1 | 4 | 656 | |||
NM_198902.2 | NP_944492.1 | 3 | 581 | |||
XM_017021857.1 | XP_016877346.1 | 4 | 265 | |||
GenBank mRNA | AF054840.1 | - | 4 | 382 | ||
AF133423.1 | - | 4 | 456 | |||
AK001305.1 | - | 4 | 502 | |||
AK001310.1 | - | 4 | 488 | |||
AK001326.1 | - | 4 | 599 | |||
AK027793.1 | - | 1 | 1707 | |||
AK075336.1 | - | 4 | 537 | |||
AK075482.1 | - | 3 | 462 | |||
AK293677.1 | - | 5 | 445 | |||
AK298157.1 | - | 3 | 345 | |||
BC000704.2 | - | 4 | 557 | 3349181 | ||
BC004280.1 | - | 4 | 518 | 3616380 | ||
BC009248.2 | - | 4 | 521 | 2964568 | ||
BC011206.1 | - | 4 | 550 | 3457158 |