Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172695.1 | NP_001166166.1 | 5 | 1043 | ||
NM_001172696.1 | NP_001166167.1 | 7 | 1194 | |||
NM_005726.5 | NP_005717.3 | 6 | 1131 | |||
GenBank mRNA | AK295323.1 | - | 8 | 1363 | ||
AK304621.1 | - | 5 | 1027 | |||
BC022862.1 | - | 7 | 1167 | 4076880 | ||
BC093068.1 | - | 6 | 1086 | 30722527 | ||
L37936.1 | - | 5 | 982 |