Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127708.2 | NP_001121180.2 | 6 | 582 | ||
NM_001127709.2 | NP_001121181.2 | 5 | 426 | |||
NM_001127710.2 | NP_001121182.2 | 4 | 303 | |||
NM_001303232.1 | NP_001290161.1 | 6 | 576 | |||
NM_005807.4 | NP_005798.3 | 7 | 705 | |||
XM_017000002.1 | XP_016855491.1 | 4 | 722 | |||
XM_017000003.1 | XP_016855492.1 | 3 | 289 | |||
GenBank mRNA | AK131434.1 | - | 5 | 566 | ||
AK297718.1 | - | 4 | 420 | |||
AK301343.1 | - | 7 | 700 | |||
AK301487.1 | - | 5 | 576 | |||
AK308235.1 | - | 3 | 297 | |||
DC295855.1 | - | 4 | 300 | |||
DC404857.1 | - | 4 | 420 | |||
U70136.1 | - | 6 | 687 |