Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PROM1 SIRNA Assays ›
Gene Symbol:
PROM1
Gene Name:
prominin 1
Gene Aliases:
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
Chromosome Location:
Chr.4: 15968226 - 16084100 on Build GRCh38
UniGene ID:
Hs.614734
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138145
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PROM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145847.1
NP_001139319.1
4
932
NM_001145848.1
NP_001139320.1
4
648
NM_001145849.1
NP_001139321.1
4
652
NM_001145850.1
NP_001139322.1
4
652
NM_001145851.1
NP_001139323.1
3
625
NM_001145852.1
NP_001139324.1
3
625
NM_006017.2
NP_006008.1
4
652
XM_005248195.4
XP_005248252.1
4
748
XM_005248196.4
XP_005248253.1
4
748
XM_006713974.3
XP_006714037.1
3
253
XM_011513890.1
XP_011512192.1
5
734
XM_011513892.2
XP_011512194.1
5
885
XM_011513893.2
XP_011512195.1
5
959
XM_011513894.2
XP_011512196.1
5
1044
XM_011513895.2
XP_011512197.1
6
1160
XM_011513896.2
XP_011512198.1
5
789
XM_011513897.2
XP_011512199.1
5
976
XM_011513898.2
XP_011512200.2
5
734
XM_011513899.2
XP_011512201.2
4
748
XM_011513900.2
XP_011512202.1
5
734
XM_011513902.2
XP_011512204.1
5
734
XM_011513903.2
XP_011512205.1
4
283
XM_017008799.1
XP_016864288.1
4
778
XM_017008800.1
XP_016864289.1
5
734
XM_017008801.1
XP_016864290.1
5
734
XM_017008802.1
XP_016864291.1
5
826
XM_017008803.1
XP_016864292.1
4
723
XM_017008804.1
XP_016864293.1
4
793
XM_017008805.1
XP_016864294.1
4
666
GenBank mRNA
AF027208.1
-
4
477
AF117225.1
-
4
693
AF507034.1
-
3
450
AK027420.1
-
4
671
AK027422.1
-
4
693
AK075411.1
-
4
648
AK226033.1
-
4
671
AK310415.1
-
4
498
AK314529.1
-
4
693
AK315889.1
-
1
81
AY449689.1
-
3
413
AY449690.1
-
3
413
AY449691.1
-
3
413
AY449692.1
-
4
440
AY449693.1
-
4
440
BC012089.1
-
4
648
4644690
HQ628626.1
-
3
413
HQ628627.1
-
5
669
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
retina layer formation
photoreceptor cell maintenance
retina morphogenesis in camera-type eye
camera-type eye photoreceptor cell differentiation
glomerular visceral epithelial cell differentiation
glomerular parietal epithelial cell differentiation
positive regulation of nephron tubule epithelial cell differentiation
protein binding
actinin binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0