Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPEPPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006310.3 | NP_006301.3 | 15, 16 | 1959 | ||
XM_011525496.1 | XP_011523798.1 | 16, 17 | 1761 | |||
XM_017025372.1 | XP_016880861.1 | 15, 16 | 1921 | |||
XM_017025373.1 | XP_016880862.1 | 16, 17 | 1761 | |||
GenBank mRNA | AJ132583.1 | - | 15, 16 | 1689 | ||
AK095877.1 | - | 16, 17 | 1725 | |||
AK096709.1 | - | 17, 18 | 1746 | |||
AK293450.1 | - | 9, 10 | 891 | |||
AK293995.1 | - | 8, 9 | 873 | |||
AK296887.1 | - | 14, 15 | 1602 | |||
AK297107.1 | - | 14, 15 | 1463 | |||
AK303037.1 | - | 16, 17 | 1754 | |||
BC065294.1 | - | 15, 16 | 1648 | 6059589 |