Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006536.5 | NP_006527.1 | 3 | 547 | ||
XM_011542448.2 | XP_011540750.1 | 3 | 489 | |||
GenBank mRNA | AB026833.1 | - | 3 | 426 | ||
AF043977.1 | - | 3 | 493 | |||
AF127980.1 | - | 3 | 385 | |||
AK290348.1 | - | 3 | 452 | |||
AK307937.1 | - | 3 | 450 | |||
BC041096.1 | - | 3 | 448 | 5752653 |