Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006778.3 | NP_006769.2 | 3 | 686 | ||
NM_052828.2 | NP_439893.2 | 3 | 686 | |||
XM_011514221.1 | XP_011512523.1 | 5 | 934 | |||
XM_011514222.2 | XP_011512524.1 | 5 | 1168 | |||
XM_011514223.2 | XP_011512525.1 | 6 | 1049 | |||
XM_011514225.1 | XP_011512527.1 | 5 | 934 | |||
GenBank mRNA | AF220122.1 | - | 3 | 684 | ||
AF220123.1 | - | 3 | 684 | |||
BC093926.1 | - | 3 | 664 | 7939771 | ||
BC113419.1 | - | 3 | 664 | 8322471 |