Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135940.1 | NP_001129412.1 | 10 | 1357 | ||
NM_001300911.1 | NP_001287840.1 | 11 | 1488 | |||
NM_006790.2 | NP_006781.1 | 10 | 1839 | |||
XM_017010060.1 | XP_016865549.1 | 11 | 2057 | |||
XM_017010061.1 | XP_016865550.1 | 9 | 1490 | |||
XM_017010062.1 | XP_016865551.1 | 10 | 1926 | |||
GenBank mRNA | AF133820.2 | - | 10 | 1824 | ||
AF144477.1 | - | 10 | 1810 | |||
AK300076.1 | - | 11 | 1468 | |||
AK300088.1 | - | 10 | 1355 | |||
BC005376.1 | - | 10 | 1819 | 3934230 |