Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013388.4 | NP_037520.1 | 1 | 379 | ||
XM_006711914.2 | XP_006711977.1 | 1 | 360 | |||
XM_011532471.2 | XP_011530773.1 | 1 | 360 | |||
XM_011532472.2 | XP_011530774.1 | 1 | 360 | |||
GenBank mRNA | AF203687.1 | - | 1 | 257 | ||
AF226684.1 | - | 1 | 270 | |||
AK023064.1 | - | 1 | 336 | |||
AK023405.1 | - | 1 | 423 | |||
AK292178.1 | - | 2 | 266 | |||
BC002765.2 | - | 1 | 310 | 3631721 | ||
BC012890.1 | - | 1 | 320 | 4158577 | ||
BC016472.1 | - | 1 | 310 | 4178911 | ||
BC041032.1 | - | 1 | 373 | 4638313 | ||
BG715668.1 | - | 1 | 385 |