Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142610.1 | NP_001136082.1 | 8 | 1069 | ||
NM_014683.3 | NP_055498.3 | 8 | 1069 | |||
XM_011524087.2 | XP_011522389.1 | 4 | 366 | |||
XM_017025424.1 | XP_016880913.1 | 8 | 957 | |||
XM_017025425.1 | XP_016880914.1 | 8 | 957 | |||
XM_017025426.1 | XP_016880915.1 | 8 | 957 | |||
XM_017025427.1 | XP_016880916.1 | 7 | 470 | |||
XM_017025428.1 | XP_016880917.1 | 7 | 470 | |||
GenBank mRNA | AB014523.1 | - | 8 | 1060 | ||
AK098316.1 | - | 7 | 694 | |||
AK314345.1 | - | 8 | 594 | |||
BC034988.1 | - | 9 | 634 | 5268025 |