Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304743.1 | NP_001291672.1 | 9 | 2270 | ||
NM_014720.3 | NP_055535.2 | 9 | 2270 | |||
GenBank mRNA | AB002804.1 | - | 9 | 1941 | ||
AF273048.1 | - | 9 | 1743 | |||
AK302855.1 | - | 9 | 1703 | |||
AK307389.1 | - | 10 | 1822 | |||
BC047762.1 | - | 9 | 2263 | |||
BC064804.1 | - | 9 | 2266 | |||
D86959.1 | - | 9 | 2220 |