Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278589.1 | NP_001265518.1 | 12 | 1545 | ||
NM_003932.4 | NP_003923.2 | 12 | 1575 | |||
GenBank mRNA | AK130770.1 | - | 12 | 1208 | ||
AK303530.1 | - | 12 | 1176 | |||
AK316155.1 | - | 12 | 1094 | |||
AY513286.1 | - | 12 | 1173 | |||
AY826824.1 | - | 12 | 1184 | |||
AY826825.1 | - | 12 | 1184 | |||
BC052982.1 | - | 12 | 1197 | 6046993 | ||
BC071629.1 | - | 12 | 1175 | 4536907 | ||
BC107148.1 | - | 10 | 824 | 40031944 | ||
BC139724.1 | - | 12 | 1132 | 40031942 | ||
CR456586.1 | - | 12 | 1215 | |||
U17714.1 | - | 12 | 1181 | |||
U28918.1 | - | 12 | 1200 |