Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105251.2 | NP_001098721.1 | 4 | 1099 | ||
NM_001284236.1 | NP_001271165.1 | 4 | 1025 | |||
NM_001284237.1 | NP_001271166.1 | 3 | 971 | |||
NM_014733.4 | NP_055548.3 | 3 | 971 | |||
XM_005248632.4 | XP_005248689.1 | 3 | 1097 | |||
XM_011543753.2 | XP_011542055.1 | 4 | 996 | |||
XM_017010091.1 | XP_016865580.1 | 5 | 1157 | |||
XM_017010092.1 | XP_016865581.1 | 4 | 1143 | |||
XM_017010095.1 | XP_016865584.1 | 4 | 996 | |||
GenBank mRNA | AB002303.1 | - | 4 | 1039 | ||
AF434817.1 | - | 2 | 791 | |||
AL833087.2 | - | 1 | 18 | |||
BC030808.1 | - | 3 | 901 | 4701661 | ||
BX537424.1 | - | 3 | 923 | |||
CR933621.1 | - | 4 | 974 |