Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KALRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024660.4 | NP_001019831.2 | 1, 2 | 232 | ||
NM_001322988.1 | NP_001309917.1 | 1, 2 | 232 | |||
NM_001322989.1 | NP_001309918.1 | 1, 2 | 232 | |||
NM_001322990.1 | NP_001309919.1 | 1, 2 | 232 | |||
NM_001322991.1 | NP_001309920.1 | 1, 2 | 232 | |||
NM_001322992.1 | NP_001309921.1 | 1, 2 | 232 | |||
NM_003947.5 | NP_003938.1 | 1, 2 | 232 | |||
XM_011513281.2 | XP_011511583.1 | 1, 2 | 23126 | |||
GenBank mRNA | AK314126.1 | - | 1, 2 | 232 | ||
U94190.1 | - | 1, 2 | 175 |