Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324310.1 | NP_001311239.1 | 2, 3 | 566 | ||
NM_001324311.1 | NP_001311240.1 | 2, 3 | 576 | |||
NM_014864.3 | NP_055679.1 | 2, 3 | 671 | |||
GenBank mRNA | AB007944.1 | - | 2, 3 | 714 | ||
AB480690.1 | - | 2, 3 | 671 | |||
AK289989.1 | - | 2, 3 | 571 | |||
BC046441.1 | - | 2, 3 | 539 | 5197154 | ||
BC051794.1 | - | 2, 3 | 671 | 5492043 |