Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRSK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256627.1 | NP_001243556.1 | 2 | 526 | ||
NM_001256629.1 | NP_001243558.1 | 2 | 526 | |||
NM_001256630.1 | NP_001243559.1 | 2 | 531 | |||
NM_001282218.1 | NP_001269147.1 | 2 | 513 | |||
NM_003957.3 | NP_003948.2 | 2 | 526 | |||
NR_046331.1 | 2 | 526 | ||||
XM_005253218.3 | XP_005253275.1 | 2 | 238 | |||
XM_006718374.2 | XP_006718437.1 | 2 | 238 | |||
XM_006718376.2 | XP_006718439.1 | 2 | 150 | |||
XM_011520461.2 | XP_011518763.1 | 2 | 150 | |||
XM_011520462.2 | XP_011518764.1 | 2 | 150 | |||
XM_017018532.1 | XP_016874021.1 | 2 | 150 | |||
XM_017018533.1 | XP_016874022.1 | 2 | 238 | |||
XM_017018534.1 | XP_016874023.1 | 2 | 150 | |||
XM_017018535.1 | XP_016874024.1 | 2 | 150 | |||
XM_017018536.1 | XP_016874025.1 | 2 | 238 | |||
XM_017018537.1 | XP_016874026.1 | 2 | 150 | |||
GenBank mRNA | AF533876.1 | - | 2 | 524 | ||
AF533877.1 | - | 2 | 524 | |||
AF533878.1 | - | 2 | 524 | |||
AF533879.1 | - | 2 | 524 | |||
AF533880.1 | - | 2 | 524 | |||
AK122851.1 | - | 2 | 515 | |||
AK131534.1 | - | 2 | 531 | |||
AY166857.1 | - | 2 | 526 | |||
AY505125.1 | - | 2 | 140 | |||
DR001379.1 | - | 2 | 244 |