Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018714.2 | NP_061184.1 | 7 | 2018 | ||
GenBank mRNA | AB037802.1 | - | 7 | 1980 | ||
AK098224.1 | - | 7 | 1738 | |||
AL359611.1 | - | 1 | 358 | |||
BC021985.2 | - | 7 | 1983 | 2821581 | ||
BC047465.1 | - | 8 | 2320 | 5285077 |