Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTPIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024430.3 | NP_077748.3 | 7 | 539 | ||
XM_011526253.2 | XP_011524555.1 | 6 | 504 | |||
XM_011526254.1 | XP_011524556.1 | 7 | 515 | |||
XM_011526256.1 | XP_011524558.1 | 6 | 415 | |||
GenBank mRNA | AK023100.1 | - | 3 | 107 | ||
BC035395.1 | - | 7 | 512 | 4524856 | ||
BP352927.1 | - | 7 | 539 | |||
KU178455.1 | - | 7 | 443 | |||
KU178456.1 | - | 7 | 443 | |||
KU178457.1 | - | 6 | 365 |