Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201572.1 | NP_001188501.1 | 4, 5 | 678 | ||
NM_001201584.1 | NP_001188513.1 | 4, 5 | 740 | |||
NM_004206.3 | NP_004197.1 | 4, 5 | 740 | |||
NM_032970.3 | NP_116752.1 | 4, 5 | 740 | |||
XM_005265533.1 | XP_005265590.1 | 5, 6 | 914 | |||
XM_005265534.1 | XP_005265591.1 | 5, 6 | 814 | |||
XM_005265535.1 | XP_005265592.1 | 5, 6 | 872 | |||
XM_011534212.1 | XP_011532514.1 | 4, 5 | 592 | |||
XM_017007438.1 | XP_016862927.1 | 5, 6 | 914 | |||
XM_017007439.1 | XP_016862928.1 | 5, 6 | 934 | |||
GenBank mRNA | AF039568.1 | - | 3, 4 | 530 | ||
AK074576.1 | - | 4, 5 | 740 | |||
AK292856.1 | - | 4, 5 | 674 | |||
AK293133.1 | - | 5, 6 | 790 | |||
AY359041.1 | - | 4, 5 | 666 | |||
BC006178.1 | - | 4, 5 | 645 | 4054516 | ||
BC018437.1 | - | 4, 5 | 648 | 3445481 | ||
BX377531.2 | - | 4, 5 | 679 | |||
CR749670.1 | - | 2, 3 | 294 | |||
KU178463.1 | - | 3, 4 | 530 | |||
KU178464.1 | - | 3, 4 | 530 | |||
KU178465.1 | - | 2, 3 | 366 |