Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000368.4 | NP_000359.1 | 3 | 311 | ||
NM_001162426.1 | NP_001155898.1 | 3 | 311 | |||
NM_001162427.1 | NP_001155899.1 | 3 | 311 | |||
XM_005272211.1 | XP_005272268.1 | 3 | 356 | |||
XM_006717271.1 | XP_006717334.1 | 3 | 291 | |||
XM_011518979.2 | XP_011517281.1 | 3 | 305 | |||
XM_017015096.1 | XP_016870585.1 | 4 | 770 | |||
XM_017015097.1 | XP_016870586.1 | 4 | 741 | |||
XM_017015098.1 | XP_016870587.1 | 3 | 356 | |||
XM_017015099.1 | XP_016870588.1 | 3 | 553 | |||
XM_017015100.1 | XP_016870589.1 | 3 | 524 | |||
XM_017015101.1 | XP_016870590.1 | 3 | 554 | |||
GenBank mRNA | AB190910.1 | - | 3 | 273 | ||
AF013168.1 | - | 3 | 298 | |||
AK297326.1 | - | 3 | 297 | |||
AK303030.1 | - | 3 | 294 | |||
AK308412.1 | - | 3 | 294 | |||
BC047772.2 | - | 3 | 284 | |||
BC070032.1 | - | 3 | 312 | |||
BC108668.1 | - | 3 | 301 | |||
BC121000.2 | - | 3 | 268 | |||
BI463204.1 | - | 3 | 317 | |||
DA726552.1 | - | 3 | 312 | |||
KJ534953.1 | - | 1 | 77 |