Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286485.1 | NP_001273414.1 | 14, 15 | 1213 | ||
NM_001286486.1 | NP_001273415.1 | 13, 14 | 1081 | |||
NM_003834.2 | NP_003825.1 | 15, 16 | 1257 | |||
NM_183337.2 | NP_899180.1 | 15, 16 | 1246 | |||
XM_011522719.2 | XP_011521021.1 | 16, 17 | 1340 | |||
XM_011522720.2 | XP_011521022.1 | 16, 17 | 1280 | |||
GenBank mRNA | AB016929.1 | - | 14, 15 | 1181 | ||
AB209582.1 | - | 7, 8 | 3154 | |||
AF035153.1 | - | 15, 16 | 1217 | |||
AF035154.1 | - | 15, 16 | 1228 | |||
AK091701.1 | - | 13, 14 | 1078 | |||
AK293564.1 | - | 15, 16 | 1127 | |||
BC141803.1 | - | 15, 16 | 1221 | 40147243 |