Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003215.2 | NP_003206.2 | 18 | 3205 | ||
XM_011513737.2 | XP_011512039.1 | 18 | 3186 | |||
XM_017008569.1 | XP_016864058.1 | 17 | 4411 | |||
GenBank mRNA | D29767.1 | - | 18 | 3231 |