Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323964.1 | NP_001310893.1 | 2, 3 | 701 | ||
NM_001323967.1 | NP_001310896.1 | 2, 3 | 701 | |||
NM_001323969.1 | NP_001310898.1 | 3, 4 | 763 | |||
NM_003252.3 | NP_003243.1 | 2, 3 | 701 | |||
NR_136661.1 | 2, 3 | 701 | ||||
NR_136663.1 | 3, 4 | 763 | ||||
GenBank mRNA | AK290695.1 | - | 2, 3 | 654 | ||
AK291239.1 | - | 2, 3 | 334 | |||
AK295243.1 | - | 2, 3 | 146 | |||
AL833106.1 | - | 2, 3 | 637 | |||
AY633610.1 | - | 2, 3 | 145 | |||
BC030025.1 | - | 2, 3 | 706 | 4821759 | ||
D64015.1 | - | 2, 3 | 177 | |||
M96954.1 | - | 2, 3 | 182 |