Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003253.2 | NP_003244.2 | 6, 7 | 1891 | ||
XM_005261037.1 | XP_005261094.1 | 5, 6 | 2024 | |||
XM_005261038.2 | XP_005261095.1 | 4, 5 | 1758 | |||
XM_005261040.2 | XP_005261097.1 | 5, 6 | 2024 | |||
XM_011529711.1 | XP_011528013.1 | 6, 7 | 1975 | |||
XM_011529712.1 | XP_011528014.1 | 6, 7 | 1899 | |||
XM_011529713.1 | XP_011528015.1 | 6, 7 | 2309 | |||
XM_017028447.1 | XP_016883936.1 | 6, 7 | 2078 | |||
XM_017028448.1 | XP_016883937.1 | 6, 7 | 2048 | |||
XM_017028449.1 | XP_016883938.1 | 5, 6 | 1955 | |||
XM_017028450.1 | XP_016883939.1 | 5, 6 | 1975 | |||
XM_017028451.1 | XP_016883940.1 | 5, 6 | 2455 | |||
XM_017028452.1 | XP_016883941.1 | 7, 8 | 2255 | |||
XM_017028453.1 | XP_016883942.1 | 5, 6 | 1930 | |||
GenBank mRNA | AB209101.1 | - | 2, 3 | 907 | ||
AK093621.1 | - | 4, 5 | 748 | |||
BC117192.1 | - | 3, 4 | 1466 | 40125743 | ||
BC117196.1 | - | 3, 4 | 1466 | 40125747 | ||
BC143970.1 | - | 3, 4 | 1466 | |||
BC143980.1 | - | 3, 4 | 1466 | 9052498 | ||
U16296.1 | - | 6, 7 | 1892 |