Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278740.1 | NP_001265669.1 | 8 | 1326 | ||
NM_001278741.1 | NP_001265670.1 | 8 | 1096 | |||
NM_001278742.1 | NP_001265671.1 | 7 | 988 | |||
NM_001278743.1 | NP_001265672.1 | 7 | 1012 | |||
NM_003270.3 | NP_003261.1 | 8 | 1034 | |||
XM_011531018.1 | XP_011529320.1 | 9 | 836 | |||
GenBank mRNA | AB097016.1 | - | 8 | 930 | ||
AB209876.1 | - | 8 | 993 | |||
AF043906.1 | - | 8 | 937 | |||
AF053453.1 | - | 8 | 890 | |||
AF133426.1 | - | 8 | 897 | |||
AK075072.1 | - | 8 | 983 | |||
AK299176.1 | - | 9 | 753 | |||
AK300852.1 | - | 9 | 974 | |||
AY358825.1 | - | 8 | 911 | |||
BC012389.1 | - | 8 | 974 | 3857537 | ||
CX868833.1 | - | 8 | 805 | |||
U84895.1 | - | 8 | 937 |