Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101669.1 | NP_001095139.1 | 7 | 822 | ||
NM_003866.2 | NP_003857.2 | 8 | 921 | |||
XM_011532391.2 | XP_011530693.1 | 7 | 906 | |||
XM_011532392.2 | XP_011530694.1 | 7 | 672 | |||
XM_017008794.1 | XP_016864283.1 | 7 | 906 | |||
XM_017008795.1 | XP_016864284.1 | 7 | 906 | |||
XM_017008796.1 | XP_016864285.1 | 7 | 906 | |||
GenBank mRNA | AK302936.1 | - | 6 | 588 | ||
AY753912.1 | - | 4 | 326 | |||
BC005274.1 | - | 7 | 597 | |||
BC072447.1 | - | 6 | 729 | |||
BC110918.1 | - | 6 | 679 | 6160633 | ||
BC133005.1 | - | 4 | 377 | 40147027 | ||
BC144255.1 | - | 4 | 377 | |||
BX649090.1 | - | 8 | 762 | |||
U96922.1 | - | 4 | 447 |