Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166691.1 | NP_001160163.1 | 2 | 224 | ||
NM_003318.4 | NP_003309.2 | 2 | 212 | |||
XM_011536099.2 | XP_011534401.1 | 2 | 186 | |||
XM_011536100.2 | XP_011534402.1 | 2 | 212 | |||
XM_017011242.1 | XP_016866731.1 | 2 | 670 | |||
GenBank mRNA | AK095599.1 | - | 2 | 684 | ||
AK292460.1 | - | 2 | 187 | |||
AK315696.1 | - | 2 | 129 | |||
AX775733.1 | - | 2 | 184 | |||
BC000633.2 | - | 2 | 175 | 3343925 | ||
BC032858.2 | - | 2 | 177 | 5270012 | ||
DA853919.1 | - | 2 | 212 |