Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003318.4 | NP_003309.2 | 11, 12 | 1376 | ||
XM_011536099.2 | XP_011534401.1 | 11, 12 | 1350 | |||
XM_017011242.1 | XP_016866731.1 | 12, 13 | 1905 | |||
GenBank mRNA | AK315696.1 | - | 11, 12 | 1293 | ||
AX775733.1 | - | 11, 12 | 1348 | |||
BC000633.2 | - | 11, 12 | 1339 | 3343925 | ||
BC032858.2 | - | 11, 12 | 1341 | 5270012 |