Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003323.2 | NP_003314.2 | 12 | 1538 | ||
XM_006723347.3 | XP_006723410.1 | 12 | 1538 | |||
XM_006723348.3 | XP_006723411.1 | 12 | 1482 | |||
XM_011527252.2 | XP_011525554.1 | 13 | 1642 | |||
XM_011527253.2 | XP_011525555.1 | 12 | 1529 | |||
XM_011527254.2 | XP_011525556.1 | 11 | 1439 | |||
XM_011527255.2 | XP_011525557.1 | 11 | 1486 | |||
XM_011527256.2 | XP_011525558.1 | 10 | 1453 | |||
GenBank mRNA | BC026070.2 | - | 12 | 1532 | 4837710 | |
U82469.1 | - | 12 | 1480 |