Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003328.2 | NP_003319.2 | 11 | 1186 | ||
XM_011513747.2 | XP_011512049.1 | 10 | 1138 | |||
XM_011513748.2 | XP_011512050.1 | 4 | 474 | |||
XM_017008580.1 | XP_016864069.1 | 12 | 2388 | |||
XM_017008581.1 | XP_016864070.1 | 10 | 2197 | |||
XM_017008582.1 | XP_016864071.1 | 5 | 621 | |||
GenBank mRNA | AK300141.1 | - | 1 | 196 | ||
AK307055.1 | - | 1 | 82 | |||
L27071.1 | - | 11 | 1186 |