Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003380.3 | NP_003371.2 | 10 | 2094 | ||
XM_006717500.1 | XP_006717563.1 | 9 | 3169 | |||
GenBank mRNA | AF328728.1 | - | 9 | 1810 | ||
AK056766.1 | - | 10 | 2167 | |||
AK091813.1 | - | 10 | 1785 | |||
AK093924.1 | - | 6 | 2625 | |||
AK097336.1 | - | 10 | 1789 | |||
AK098444.1 | - | 10 | 1664 | |||
AK290643.1 | - | 9 | 1706 | |||
BC000163.2 | - | 9 | 1805 | 2985712 | ||
BC066956.1 | - | 10 | 1882 | 4823475 | ||
M25246.1 | - | 9 | 1185 | |||
X56134.1 | - | 9 | 1724 | |||
Z19554.2 | - | 9 | 1808 |