Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032409.2 | NP_115785.1 | 8 | 2534 | ||
GenBank mRNA | AB053323.1 | - | 8 | 2534 | ||
AK075225.1 | - | 4 | 1533 |