Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256523.1 | NP_001243452.1 | 4 | 2671 | ||
NM_001256524.1 | NP_001243453.1 | 6 | 2529 | |||
NM_001256525.1 | NP_001243454.1 | 5 | 2429 | |||
NM_003426.3 | NP_003417.2 | 5 | 2757 | |||
NR_046282.1 | 6 | 2853 | ||||
GenBank mRNA | AB209663.1 | - | 6 | 2565 | ||
AK128267.1 | - | 2 | 3762 | |||
AK291358.1 | - | 6 | 2455 | |||
BC013395.2 | - | 5 | 2355 | 3028001 | ||
BC048295.1 | - | 5 | 2342 | 5242841 | ||
X71623.1 | - | 4 | 2189 | |||
X92715.1 | - | 4 | 3355 |