Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134709.1 | NP_001128181.1 | 5 | 607 | ||
NM_003472.3 | NP_003463.1 | 6 | 709 | |||
XM_017011273.1 | XP_016866762.1 | 6 | 681 | |||
GenBank mRNA | AK294079.1 | - | 6 | 592 | ||
AK297749.1 | - | 5 | 580 | |||
AK298089.1 | - | 6 | 621 | |||
AK312616.1 | - | 6 | 638 | |||
BC035259.1 | - | 6 | 681 | 5122743 | ||
X64229.1 | - | 6 | 549 |