Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003498.5 | NP_003489.1 | 2 | 763 | ||
XM_017023741.1 | XP_016879230.1 | 2 | 791 | |||
XM_017023742.1 | XP_016879231.1 | 2 | 689 | |||
GenBank mRNA | AF030196.1 | - | 2 | 715 | ||
AF070673.1 | - | 2 | 745 | |||
AL161976.1 | - | 2 | 724 | |||
AL538273.3 | - | 2 | 762 | |||
BC036100.1 | - | 2 | 741 | 5277179 | ||
BC036443.1 | - | 2 | 735 | 4823944 |