Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134419.1 | NP_001127891.1 | 7 | 743 | ||
NM_001134420.1 | NP_001127892.1 | 7 | 871 | |||
NM_003503.3 | NP_003494.1 | 7 | 777 | |||
XM_005271241.2 | XP_005271298.1 | 7 | 739 | |||
XM_005271244.2 | XP_005271301.1 | 7 | 777 | |||
XM_017002425.1 | XP_016857914.1 | 7 | 739 | |||
XM_017002426.1 | XP_016857915.1 | 7 | 722 | |||
XM_017002427.1 | XP_016857916.1 | 6 | 585 | |||
GenBank mRNA | AB003698.1 | - | 7 | 744 | ||
AB209337.1 | - | 6 | 1453 | |||
AF005209.1 | - | 6 | 612 | |||
AF015592.1 | - | 7 | 757 | |||
AK298963.1 | - | 6 | 689 | |||
AK312726.1 | - | 7 | 833 | |||
BC110526.1 | - | 7 | 700 | 40034178 | ||
BC110527.1 | - | 7 | 734 | 40034179 | ||
BC111044.1 | - | 7 | 757 | 5518616 |