Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003592.2 | NP_003583.2 | 16 | 2120 | ||
XM_005250060.4 | XP_005250117.1 | 16 | 2156 | |||
XM_011516629.2 | XP_011514931.1 | 16 | 2112 | |||
XM_011516630.2 | XP_011514932.1 | 16 | 2013 | |||
XM_011516631.2 | XP_011514933.1 | 16 | 2141 | |||
XM_011516632.2 | XP_011514934.1 | 16 | 2039 | |||
XM_017012723.1 | XP_016868212.1 | 15 | 1876 | |||
GenBank mRNA | AF062536.1 | - | 15 | 1894 | ||
AK096163.1 | - | 17 | 2008 | |||
AK096505.1 | - | 9 | 1777 | |||
AK296909.1 | - | 15 | 1845 | |||
AK315023.1 | - | 16 | 2296 | |||
BC125119.1 | - | 16 | 2033 | 40118923 | ||
BC125120.1 | - | 16 | 2033 | 40118927 | ||
BX537409.1 | - | 16 | 2050 | |||
U58087.1 | - | 16 | 1822 |