Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323303.1 | NP_001310232.1 | 4 | 588 | ||
NM_001323304.1 | NP_001310233.1 | 4 | 351 | |||
NM_001323305.1 | NP_001310234.1 | 5 | 461 | |||
NM_003600.3 | NP_003591.2 | 3 | 478 | |||
NM_198433.2 | NP_940835.1 | 5 | 686 | |||
NM_198434.2 | NP_940836.1 | 4 | 377 | |||
NM_198435.2 | NP_940837.1 | 3 | 267 | |||
NM_198436.2 | NP_940838.1 | 4 | 363 | |||
NM_198437.2 | NP_940839.1 | 3 | 253 | |||
XM_017028034.1 | XP_016883523.1 | 5 | 808 | |||
XM_017028035.1 | XP_016883524.1 | 3 | 168 | |||
GenBank mRNA | AF008551.1 | - | 2 | 78 | ||
AF011468.1 | - | 3 | 334 | |||
AK301769.1 | - | 3 | 144 | |||
AK312776.1 | - | 3 | 145 | |||
AL711075.1 | - | 3 | 346 | |||
BC001280.1 | - | 4 | 210 | 3456017 | ||
BC002499.2 | - | 3 | 206 | 3051177 | ||
BC006423.1 | - | 3 | 103 | 3532438 | ||
BC027464.1 | - | 4 | 245 | 4449474 | ||
BF029182.1 | - | 134 | ||||
BM458030.1 | - | 4 | 278 | |||
BP224535.1 | - | 3 | 267 | |||
BQ435059.1 | - | 3 | 99 | |||
BU171303.1 | - | 3 | 199 | |||
BU542363.1 | - | 5 | 538 | |||
CB113289.1 | - | 5 | 380 | |||
CB138172.1 | - | 4 | 320 | |||
D84212.1 | - | 4 | 247 |