Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009182.1 | NP_001009182.1 | 8 | 749 | ||
NM_003616.2 | NP_003607.1 | 9 | 794 | |||
GenBank mRNA | AB037701.1 | - | 8 | 727 | ||
AB037703.1 | - | 9 | 692 | |||
AF027150.1 | - | 9 | 855 | |||
AK313947.1 | - | 9 | 782 | |||
BC028095.1 | - | 10 | 911 | |||
BC104968.1 | - | 9 | 793 | 8143971 |