Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195303.2 | NP_001182232.1 | 3 | 1281 | ||
NM_001254748.1 | NP_001241677.1 | 6 | 1339 | |||
NM_001254749.1 | NP_001241678.1 | 5 | 1466 | |||
NM_003617.3 | NP_003608.1 | 5 | 1454 | |||
GenBank mRNA | AB008109.1 | - | 5 | 1197 | ||
AF159570.1 | - | 5 | 1160 | |||
BC030059.1 | - | 5 | 1179 | 4098248 | ||
BX537427.1 | - | 5 | 1198 |