Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIBP1 SIRNA Assays ›
Gene Symbol:
PPFIBP1
Gene Name:
PPFIA binding protein 1
Gene Aliases:
L2, SGT2, hSGT2, hSgt2p
Chromosome Location:
Chr.12: 27524112 - 27695564 on Build GRCh38
UniGene ID:
Hs.172445
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139238
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198915.1
NP_001185844.1
3
369
NM_001198916.1
NP_001185845.1
5
674
NM_003622.3
NP_003613.3
5
674
NM_177444.2
NP_803193.2
5
674
XM_005253505.1
XP_005253562.1
4
377
XM_005253507.1
XP_005253564.1
4
377
XM_005253508.1
XP_005253565.1
4
377
XM_005253509.1
XP_005253566.1
4
377
XM_006719159.3
XP_006719222.1
3
331
XM_017020056.1
XP_016875545.1
6
693
XM_017020057.1
XP_016875546.1
4
868
XM_017020058.1
XP_016875547.1
4
377
XM_017020059.1
XP_016875548.1
5
660
XM_017020060.1
XP_016875549.1
4
377
XM_017020061.1
XP_016875550.1
5
643
XM_017020062.1
XP_016875551.1
5
657
XM_017020063.1
XP_016875552.1
5
660
XM_017020064.1
XP_016875553.1
5
643
XM_017020065.1
XP_016875554.1
5
643
XM_017020066.1
XP_016875555.1
5
643
XM_017020067.1
XP_016875556.1
5
643
XM_017020068.1
XP_016875557.1
5
659
XM_017020069.1
XP_016875558.1
5
643
XM_017020070.1
XP_016875559.1
5
643
XM_017020071.1
XP_016875560.1
5
643
XM_017020072.1
XP_016875561.1
5
643
XM_017020073.1
XP_016875562.1
5
660
XM_017020074.1
XP_016875563.1
3
351
XM_017020075.1
XP_016875564.1
3
309
XM_017020077.1
XP_016875566.1
4
377
XM_017020078.1
XP_016875567.1
5
648
GenBank mRNA
AB033056.2
-
3
299
AB049149.1
-
5
566
AF034802.1
-
5
537
AJ276680.1
-
4
405
AK294267.1
-
3
308
AK314350.1
-
5
562
BC022873.1
-
5
645
BC046159.1
-
5
491
BC050281.1
-
5
580
5276255
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
cell-cell adhesion
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0