Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC43A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198810.1 | NP_001185739.1 | 14 | 1611 | ||
NM_003627.5 | NP_003618.1 | 14 | 1782 | |||
XM_005274358.4 | XP_005274415.1 | 15 | 1922 | |||
XM_011545320.2 | XP_011543622.1 | 14 | 1831 | |||
XM_011545321.2 | XP_011543623.1 | 12 | 1398 | |||
XM_011545322.1 | XP_011543624.1 | 11 | 1247 | |||
XM_017018451.1 | XP_016873940.1 | 15 | 1810 | |||
XM_017018452.1 | XP_016873941.1 | 13 | 1751 | |||
XM_017018453.1 | XP_016873942.1 | 12 | 1588 | |||
GenBank mRNA | AB103033.1 | - | 14 | 1749 | ||
AF045584.1 | - | 14 | 1569 | |||
AK027732.1 | - | 12 | 1347 | |||
BC001639.2 | - | 14 | 1611 | 2959372 |