Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271769.1 | NP_001258698.1 | 16 | 1961 | ||
NM_003664.4 | NP_003655.3 | 16 | 1949 | |||
XM_005248618.3 | XP_005248675.1 | 16 | 1921 | |||
XM_005248619.4 | XP_005248676.1 | 16 | 1921 | |||
XM_017010001.1 | XP_016865490.1 | 16 | 1701 | |||
GenBank mRNA | AK225908.1 | - | 16 | 1825 | ||
AK291201.1 | - | 16 | 1890 | |||
AK303300.1 | - | 14 | 1569 | |||
AY623424.1 | - | 16 | 1851 | |||
BC038444.1 | - | 16 | 1910 | 3914400 | ||
BX538041.1 | - | 16 | 1899 | |||
KU178426.1 | - | 16 | 1773 | |||
KU178427.1 | - | 16 | 1773 | |||
U81504.1 | - | 16 | 1865 | |||
U91931.1 | - | 16 | 1826 |