Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003675.3 | NP_003666.1 | 9, 10 | 1089 | ||
XM_011519754.2 | XP_011518056.1 | 10, 11 | 1214 | |||
XM_017016860.1 | XP_016872349.1 | 8, 9 | 1076 | |||
GenBank mRNA | AF087997.1 | - | 2, 3 | 195 | ||
AK314814.1 | - | 9, 10 | 1064 | |||
BC000794.1 | - | 9, 10 | 1011 | 3451639 | ||
U51990.1 | - | 9, 10 | 1013 |